Learn More
United Scientific Supplies Semicircular Refraction Cell
Catalog No. S23919
Change view
Click to view available options
Diameter (Metric) Lens:
12 cm
Catalog No. Diameter (Metric) Lens
S23919 12 cm
1 options
Catalog No. S23919 Supplier United Scientific Supplies Supplier No. RCSC01
Show refraction and internal reflection in water. The United Scientific Supplies Semicircular Refraction Cell is used to measure refractive index, and has uniform wall thickness to eliminate distortions.
  • Excellent for showing refraction and internal reflection in water, and to measure its refractive index
  • Has uniform wall thickness to eliminate distortions
  • 12cm diameter x 2.5cm high
Specifications
Product Type Refraction Cell
Lens Material Acrylic
For Use With (Application) Showing refraction and internal reflection in water
Diameter (Metric) Lens 12 cm
Height (Metric) 2.5 cm