Learn More
Starna Cells Inc SEMI MICRO QRTZ CEL 10MM W/STP
Catalog No. NC9056438
Encompass
Semi-Micro Cells, With PTFE Stopper, Path Length: 10mm, Polished Windows: 2, Nominal volume: 1.400 mL, Window material: Spectrosil Quartz, Useable Range: 170 to 2700nm, Cell top: PTFE stopper in ground type joint
Catalog No. NC9056438 Supplier Starna Cells Inc Supplier No. 29Q10
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Semi-Micro Cells, With PTFE Stopper, Path Length: 10mm, Polished Windows: 2, Nominal volume: 1.400 mL, Window material: Spectrosil Quartz, Useable Range: 170 to 2700nm, Cell top: PTFE stopper in ground type joint