Recombinant Protein;Human Semaphorin 4A / SEMA4A Protein;100UG;Human SEMA4A, His Tag (SEA-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 33 - His 683 (Accession # AAH20974).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.7 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human Semaphorin 4A / SEMA4A Protein;100UG;Human SEMA4A, His Tag (SEA-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 33 - His 683 (Accession # AAH20974).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.7 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.