Learn More
Sellstrom™ Chemical Splash Goggles
Built for comfortable protection
Supplier: Sellstrom™ S88210
Description
- Clear UV-absorbing polycarbonate lens
- Soft vinyl goggle frame conforms to facial contours for a secure fit
- Resists mosts chemicals
- Four indirect vents for airflow
- Mini size fit narrow or small faces
Order Info
Available in standard or fog-free version.
Specifications
| Anti-fog | |
| Chemical Splash Protection Goggles | |
| Vinyl |
| Clear | |
| Polycarbonate |