Learn More
Sellstrom™ Chemical Splash Goggles
Catalog No. 172881A
Encompass
Change view
Click to view available options
Lens Coating:
Anti-fog
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating
172881A -
172881B Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 172881A Supplier Sellstrom™ Supplier No. S88200
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Built for comfortable protection
  • Clear UV-absorbing polycarbonate lens
  • Soft vinyl goggle frame conforms to facial contours for a secure fit
  • Resists mosts chemicals
  • Four indirect vents for airflow
  • Mini size fit narrow or small faces
Order Info
Available in standard or fog-free version.
Specifications
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Frame Color Green
Material Polycarbonate
Frame Material Vinyl