Learn More
Sellstrom™ 380/390 Series Faceshields and Replacement Parts
Catalog No. 17289B
Change view
Click to view available options
Certifications/Compliance:
CE Certified
Includes:
Rachet Headgear
Product Type:
Faceshield
Replacement Window
For Use With (Equipment):
380/390 Series Faceshields
390 series faceshields
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Includes Product Type For Use With (Equipment)
17289B CE Certified Rachet Headgear Faceshield 380/390 Series Faceshields
172891A - - Replacement Window 390 series faceshields
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17289B Supplier Sellstrom™ Supplier No. S39100
Only null left
Single crown style provides low cost and high quality
  • Chemical-splash resistant
  • Extreme impact strength
  • Ratchet headgear suspension allows user to adjust size and tighten at the turn of a knob
Models and Replacements
  • 17-289-B: Blue Plastic Crown with Rachet headgear (CE Certified)
  • 19-043-978: Blue Plastic Crown with Rachet Headgear and Clear Window
  • 17-289-1A: Window: 8 x 12 x 0.040 in.
  • 19-065-757: Window: 8 x 16 x 0.040 in. (CE Certified)
Compliance
17-289-B: Meets ANSI Z87.1+ High Impact Standards, CUL certified to meet CAN/CSA Z94.3 Standards, CE certified; 19-065-757: CE certified
Specifications
Certifications/Compliance CE Certified
Product Type Faceshield
For Use With (Equipment) 380/390 Series Faceshields
Frame Color Blue
Includes Rachet Headgear
Material Plastic