missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Self Sticking Vinyl Arrows, Single Direction

Catalog No. 19111914
Change view
Click to view available options
Size (English):
12 x 4 in. (W x H)
7 x 1.25 in. (W x H)
7 x 2.25 in. (W x H)
7 x 4 in. (W x H)
Color:
Black, Orange
Black, White
Black, Yellow
White, Blue
White, Green
White, Red
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Color
19111914 12 x 4 in. (W x H) White, Blue
19111870 7 x 2.25 in. (W x H) White, Blue
19111871 7 x 2.25 in. (W x H) White, Green
19111873 7 x 2.25 in. (W x H) White, Red
19111872 7 x 2.25 in. (W x H) Black, Orange
19111875 7 x 2.25 in. (W x H) Black, Yellow
19111874 7 x 4 in. (W x H) Black, White
19111909 7 x 1.25 in. (W x H) Black, White
19111912 12 x 4 in. (W x H) Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19111914 Supplier Brady™ Supplier No. 93392

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

To identify pipe content flow. Brady Self Sticking Vinyl Arrows, Single Direction are made of vinyl designed to withstand indoor and outdoor environments.

  • Portable card-mounted
  • Available in three different sizes and various colors to fit to various pipe diameters
  • Meet ASME (ANSI) standards

Specifications

Description Arrow style: Single
Adhesive Type Acrylic, Permanent, Cold-temperature
Color White, Blue
Format Die-cut
Material Pressure-sensitive Vinyl (B-946)
Product Type Arrow Tape
For Use With (Application) Pipe and valve marking
Size (English) 12 x 4 in. (W x H)
Size (Metric) 30.48 x 10.16 cm (W x H)
Thickness (English) 0.004 in.