missing translation for 'onlineSavingsMsg'
Learn More

Slice™ Self-Opening Scissors

Catalog No. 18222836
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18222836 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18222836 Supplier Slice™ Supplier No. DTC105955

Ideal for industrial tailors and fibre artists, or anyone with arthritis, hand weakness, or dexterity challenges

  • Zirconium oxide Slice™ safety blades ideal for thread and fibres
  • Durable reinforced nylon handles with ribbed grips
  • Safer than traditional scissors
  • Self-open design lowers muscle effort
  • Finger-friendly™ edge lasts 11x longer than metal
  • Ambidextrous design
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic blades
  • Chemically inert blades never rust
  • Oil, lubricant, and maintenance free
  • Easy to clean
  • BPA, phthalate, and lead-free scissors
  • LIghtweight and compact
  • Features lanyard hole and safety cap

Specifications

Product Type Scissors
Material PA6 GF50, Polypropylene,Stainless Steel, Zirconium Oxide
Color Gray
For Use With (Application) Safety Cutting
Includes Ceramic blades
No. per Pack 1 Ea.