Learn More
Brady™ Self-laminating Vinyl Wrap Around Labels
Mark cables and wires with the B-427 self-laminating vinyl label material, designed with a clear, self-laminating protective layer. Print almost anywhere with a durable Brady™ portable label printer.
Supplier:  Brady™ M4207RO427
 View more versions of this product
Catalog No. 17-012-169
Only null left
Explore available promotions
Description
Description
  • Designed for tightly-curved surfaces such as wires and cables
  • Self-extinguishing and resistant to abrasion, solvents, oil, grease, weather and extreme temperatures
  • Designed to spin freely, allowing you to read and identify cables and wires from any angle
  • Made with a clear, non-printable overlaminate for increased protection
Specifications
Specifications
UL Recognised to UL969 Labelling and Marking Standard when printed with the Brady Series R4300 and R6200 ribbons. See UL files MH17154 for specific details. Brady customers should specify at purchase if they require UL Recognised B-427 labels. Brady sells both UL Recognised B-427 labels and non-approved labels depending on customer requirements. Compliant with Directive 2011/65/EU and 2015/863
White, Transparent
Die-cut
Label
Data and telecommunications labelling;Electrical labelling;Wire and cable labelling
70
-70°C to 70°C
0.102 mm
12.70 mm (W) x 69.85 mm (H)
0.5 in
12.70 mm
69.85 mm
Permanent Acrylic
Matte
Vinyl
Non-sterile
BMP41;BMP51;M511;M410;M510
-94°F to 158°F
0.004 in
10 AWG - 3 AWG
2.75 in.
12.70 mm
2.75 in
M4-207-RO-427
Product Suggestions
Product Suggestions