Learn More
Brady™ Self-laminating Cryogenic Polyester Labels
Securely label lab tubes and vials for cryogenic storage with this self-laminating, ultra-thin polyester label material (B-461). Print almost anywhere with a compact Brady™ portable label printer.
Supplier:  Brady™ M5122461
 View more versions of this product
Catalog No. 17012264
Only null left
Description
Description
  • Ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws and slides
  • Able to withstand extreme temperatures from -196°C to 130°C (-320°F to 266°F), immersion in liquid nitrogen and autoclave use
  • Resistant to ethanol, toluene, isopropanol, xylene, DMSO, 50% acetic acid, 10% hydrochloric acid, 10% Clorox solution
  • Made with a clear overlaminate that protects the print and maintains content visibility
Specifications
Chemical Identifiers
Autoclavable
Matte
Thermal Transfer
-321°F to +230°F
Permanent Acrylic
Polyester
Non-sterile
Specifications
Autoclavable
Permanent Acrylic
Matte
Polyester
Label
Laboratory labelling
-196°C
360
-321°F to +230°F
0.0019 in
28.58 mm (W) x 12.70 mm (H)
1.125 in
0.5 in
12.70 mm
2011/65/EU, 2015/863
White, Transparent
Die-cut
Thermal Transfer
Non-sterile
BMP51;BMP53;M511;M510
360 Label(s) / Box
28.58 mm (W) x 12.70 mm (H)
-196°C to +110°C
0.048 mm
0.5 in.
28.58 mm
0.5 in
M5-122-461
Product Suggestions
Product Suggestions