missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Self-Adhesive Document Holders

Catalog No. 19113408
Change view
Click to view available options
Dimensions (L x W):
11.5 x 9 in
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19113408 11.5 x 9 in
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19113408 Supplier Brady™ Supplier No. 112633

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Display charts, graphs and spreadsheets from your computer. The Brady™ Self-Adhesive Document Holders are easily applied to Lean Communication Boards.

  • Self-adhesive document holders
  • Made for use with Lean Communication Boards.
  • Adhesive backing allows for easy application to display charts, graphs and spreadsheets created on your computer

Specifications

Color Clear
Mounting Type Adhesive
For Use With (Equipment) Lean Communication Boards
Quantity 10/Pk.
Device Holds Charts, graphs, spreadsheets and more
Dimensions (L x W) 11.5 x 9 in
Length (English) 11.5 in.