missing translation for 'onlineSavingsMsg'
Learn More

Selenium(IV) oxide, Puratronic™, 99.999% (metals basis)

Catalog No. AA1085414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1085414 25 g
AA1085406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1085414 Supplier Thermo Scientific Chemicals Supplier No. 01085414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7446-08-4 | O2Se | 110.97 g/mol

In organic reactions, selenium dioxide is often used as an oxidizing reagent employed for carrying out allylic oxidations, Riley oxidation, synthesis of diones, etc. It is used to impart a red color to glass. It finds use as a toner in photographic developing, and an active ingredient in some cold-blueing solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7446-08-4
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
Synonym selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
PubChem CID 24007
IUPAC Name selenium dioxide
SMILES O=[Se]=O

Specifications

Melting Point 340°C to 350°C (sublimation)
Physical Form Lump/Powder
Quantity 25 g
Odor Odorless
UN Number UN3283
Merck Index 14,8434
Solubility Information Soluble in water,benzene,ethanol and methanol. Sprangly soluble in acetone and acetic acid.
Formula Weight 110.96
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Selenium(IV) oxide
Show More Show Less
Hazard Category H301-H310+H330-H314-H341-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P261-P262-P264b-P270-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
EINECSNumber 231-194-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only