missing translation for 'onlineSavingsMsg'
Learn More

Selenium(IV) bromide, 99+%

Catalog No. AA4024014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4024014 25 g
AA4024006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4024014 Supplier Thermo Scientific Chemicals Supplier No. 04024014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7789-65-3 | Br4Se | 398.587 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-65-3
Molecular Formula Br4Se
Molecular Weight (g/mol) 398.587
MDL Number MFCD00049766
InChI Key VTQZBGAODFEJOW-UHFFFAOYSA-N
Synonym selenium tetrabromide, selenium iv bromide, selenium bromide sebr4 , t-4, unii-8o54yxr180, tetrabromo-, e4-selane, sebr4, tetrabromo-$l^ 4-selane, tetrabromo-??-selane, selenium iv bromide 5g
PubChem CID 82246
SMILES [Se](Br)(Br)(Br)Br

Specifications

Melting Point 70°C to 80°C (decomposition)
Physical Form Powder
Quantity 25 g
Sensitivity Moisture sensitive
UN Number UN2923
Merck Index 14,8437
Solubility Information Decomposes in water; Soluble in carbon disulfide, chloroform, ethyl bromide
Formula Weight 398.6
Percent Purity ≥99%
Chemical Name or Material Selenium(IV) bromide
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 232-181-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only