missing translation for 'onlineSavingsMsg'
Learn More

Selenium Dioxide, Purified, Approx. 95%, Spectrum™ Chemical

Catalog No. 18613092
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613092 100 g Amber Glass Bottle
18613095 500 g Amber Glass Bottle
18613096 2.5 kg Amber Glass Bottle
18613093 12 kg Poly Pail
18613094 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613092 Supplier Spectrum Chemical Mfg Cor Supplier No. S1122100GM

Chemical Identifiers

CAS 7446-08-4

Specifications

CAS Max % 1
Quantity 100 g
Percent Purity ∼95%
Grade Purified
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.