missing translation for 'onlineSavingsMsg'
Learn More

Selenium dichloride oxide, 99% (metals basis)

CAS: 7791-23-3 | Cl2OSe | 165.87 g/mol

$635.51 - $1362.82

Chemical Identifiers

CAS 7791-23-3
Molecular Formula Cl2OSe
Molecular Weight (g/mol) 165.87
MDL Number MFCD00016327
InChI Key LIRMPIXWGWLNND-UHFFFAOYSA-N
Synonym selenium oxychloride, seleninyl chloride, selenium oxydichloride, seleninyl dichloride, selenium chloride oxide, hsdb 678, selenium chloride oxide secl2o, selenium iv oxychloride, selenium dichloride oxide, selenoxychlorid
PubChem CID 24647
SMILES O=[Se](Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA68108AB
View Documents
Thermo Scientific Chemicals
068108AB
10 mL
Each for $635.51
Only null left
 
AA68108AC
View Documents
Thermo Scientific Chemicals
068108AC
25 mL
Each for $1,362.82
Only null left
 
Description

Description

Selenium(IV) chloride oxide is used as a polar solvent for synthetic phenolic resins. It is also used to prepare selenium oxybromide and selenium nitride. It acts as a chlorinating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7791-23-3
165.87
LIRMPIXWGWLNND-UHFFFAOYSA-N
24647
Cl2OSe
MFCD00016327
selenium oxychloride, seleninyl chloride, selenium oxydichloride, seleninyl dichloride, selenium chloride oxide, hsdb 678, selenium chloride oxide secl2o, selenium iv oxychloride, selenium dichloride oxide, selenoxychlorid
O=[Se](Cl)Cl

Specifications

176.4°C
7791-23-3
Liquid
Cl2OSe
UN2879
selenium oxychloride, seleninyl chloride, selenium oxydichloride, seleninyl dichloride, selenium chloride oxide, hsdb 678, selenium chloride oxide secl2o, selenium iv oxychloride, selenium dichloride oxide, selenoxychlorid
LIRMPIXWGWLNND-UHFFFAOYSA-N
165.87
165.87
1.651
2.42 g/mL
(metals basis)
8.5°C
Yellow
10 mL
MFCD00016327
14,8435
Miscible with chloroform,carbon disulphide and benzene.
O=[Se](Cl)Cl
24647
99%
Moisture sensitive
Selenium dichloride oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P301+P310-P314-P330-P501c

H301-H373

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: SELENIUM OXYCHLORIDE

EINECSNumber : 232-244-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only