missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Tridodecylmethylammonium chloride, ≥97.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111015334
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015334 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015334 Supplier Merck Emd Millipore Supplier No. 916611G

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00043303
Synonym Methyltridodecylammonium chloride

Specifications

Melting Point 110°C to 112°C
CAS 7173-54-8
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula [CH3(CH2)11]3 N(Cl)CH3
Quantity 1 g
Formula Weight 572.47
Percent Purity ≥97.0% (TLC)
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH