missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Tetradodecylammonium nitrate, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111019496
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019496 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019496 Supplier Merck Emd Millipore Supplier No. 87252100MG
Only null left

Tetradodecylammonium nitrate (TDDAN) is a quaternary ammonium salt, mainly used as electrolyte.

Chemical Identifiers

MDL Number MFCD00467971

Specifications

Melting Point 103°C to 105°C
CAS 63893-35-6
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula [CH3(CH2)10 CH2]4 N(NO3)
Quantity 100 mg
Formula Weight 753.32
Percent Purity ≥99.0% (NT); ≥99.0%
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH