missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, ≥98.0% (based on Na, AAS), MilliporeSigma™ Supelco™

Catalog No. 111015553
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015553 50 mg
111015551 10 mg
111015552 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111015553 Supplier Merck Emd Millipore Supplier No. 7201750MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic ion which is insoluble in water. It is also strong against acids and oxidants. It can be prepared using boron trifluoride and 3,5-bis(trifluoromethyl)phenylmagnesium iodide as major components.

Chemical Identifiers

MDL Number MFCD00043323
Synonym NaBARF; Tetrakis[3,5-bis(trifluoromethyl)phenyl]boron sodium

Specifications

CAS 79060-88-1
Quantity 50 mg
Linear Formula C32H12 BF24 Na
Molecular Weight (g/mol) 886.2
Formula Weight 886.20
Percent Purity ≥98.0% (based on Na, AAS)
Grade Selectophore
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line Selectophore
Chemical Name or Material Sodium Tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH