missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Sodium tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]borate trihydrate, MilliporeSigma™ Supelco™

Catalog No. 111012757
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012757 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012757 Supplier Merck Emd Millipore Supplier No. 7201550MG

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD01863091
Synonym HFPB; Tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]boron sodium

Specifications

CAS 120945-63-3
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C56H36 BF48 NaO8 · 3H2 O
Quantity 50 mg
Formula Weight 1836.65
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH