missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Sodium ionophore X, Function Tested, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 97600-39-0 |
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| Linear Formula | C60H80O12 |
| Quantity | 50 mg |
| Formula Weight | 993.27 |
| Grade | Selectophore |
| Physical Form | Crystals |
| Product Line | Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH