missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Sodium ionophore VI, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111015593
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015593 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015593 Supplier Merck Emd Millipore Supplier No. 71739500MG
Only null left

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00010527
Synonym Bis[(12-crown-4)methyl] dodecylmethylmalonate; Dodecylmethylmalonic acid bis[(12-crown-4)methyl ester]

Specifications

CAS 80403-59-4
Flash Point 113°C - Closed Cup
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C34H62O12
Quantity 500 mg
Formula Weight 662.85
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH