missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Silanization solution I ∼5 (dimethyldichlorosilane in heptane), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 75-78-5 |
| Quantity | 250 mL |
| Linear Formula | C2H6 Cl2 Si |
| Flash Point | -4°C |
| Formula Weight | 129.06 |
| Percent Purity | 5% (dimethyldichlorosilane in heptane) |
| Grade | Selectophore |
| Product Line | Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH