missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Silanization solution I ∼5 (dimethyldichlorosilane in heptane), MilliporeSigma™ Supelco™

Catalog No. 111019366
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019366 250 mL
111018310 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019366 Supplier Merck Emd Millipore Supplier No. 85126250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00000491
Synonym Dichlorodimethylsilane solution; Dimethyldichlorosilane solution

Specifications

CAS 75-78-5
Quantity 250 mL
Linear Formula C2H6 Cl2 Si
Flash Point -4°C
Formula Weight 129.06
Percent Purity 5% (dimethyldichlorosilane in heptane)
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH