missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Potassium ionophore I, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111018150
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018150 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018150 Supplier Merck Emd Millipore Supplier No. 60403100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

CAS 2001-95-8
Molecular Weight (g/mol) 1111.32
MDL Number MFCD00005114
Synonym Valinomycin

Specifications

Melting Point 184°C to 190°C
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C54H90N6O18
Quantity 100 mg
Formula Weight 1111.32
Percent Purity ≥90.0%
Grade Selectophore
Physical Form Powder
Product Line Selectophore
RTECSNumber YV9468000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH