missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Monensin methyl ester (TLC), ≥97.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111018707
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018707 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018707 Supplier Merck Emd Millipore Supplier No. 30552100MG
Only null left

Chemical Identifiers

Synonym Methyl monensin

Specifications

CAS 28636-21-7
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C37H64O11
Quantity 100 mg
Formula Weight 684.90
Percent Purity ≥97.0% (TLC)
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH