missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Lead Ionophore IV, MilliporeSigma™ Supelco™

Catalog No. 111013134
Change view
Click to view available options
Quantity:
50 mg
Packaging:
Bottomless Glass Bottle with Fused Cone
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111013134 50 mg Bottomless Glass Bottle with Fused Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013134 Supplier Merck Emd Millipore Supplier No. 1534350MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ionophore giving highly Pb-selective polymer membrane electrodes

Chemical Identifiers

CAS 145237-46-3
Molecular Formula C60H84N4O4S4
Molecular Weight (g/mol) 1053.60
MDL Number MFCD00679123
InChI Key CDVPPFVKRDRQTO-UHFFFAOYSA-N
Synonym tert-Butylcalix[4 ]arene-tetrakis(N,N-dimethylthioacetamide)
IUPAC Name N,N-dimethyl-2-({5,11,17,23-tetra-tert-butyl-26,27,28-tris[(dimethylcarbamothioyl)methoxy]pentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-25-yl}oxy)ethanethioamide
SMILES CN(C)C(=S)COC1=C2CC3=CC(=CC(CC4=C(OCC(=S)N(C)C)C(CC5=C(OCC(=S)N(C)C)C(CC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3OCC(=S)N(C)C)C(C)(C)C

Specifications

Quantity 50 mg
Linear Formula C60H84N4O4S4
Formula Weight 1053.59
Grade Selectophore
Packaging Bottomless Glass Bottle with Fused Cone
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH