missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Dichloromethane, Inhibitor-free, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018705
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018705 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018705 Supplier Merck Emd Millipore Supplier No. 91301500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dichloromethane is a volatile halogenated organic solvent used as a degreaser, extraction medium, lacquer remover, or paint stripper.

Chemical Identifiers

MDL Number MFCD00000881
Synonym Methylene chloride

Specifications

Melting Point -97°C
CAS 75-09-2
Density 1.325 g/mL (at 25°C)
Quantity 500 mL
Ignition Point 1223°F
Product Line Selectophore
Vapor Pressure 24.45 psi (55°C)
Linear Formula CH2 Cl2
Boiling Point 39.8°C to 40°C
Percent Purity ≥99.5% (GC)
Grade Selectophore
Show More Show Less
RTECSNumber PA8050000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH