missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Cyclohexanone, For ion-selective electrodes, ≥99.5%, MilliporeSigma™ Supelco™
Cyclohexanone is an industrially important reagent. It is also a major intermediate in the synthesis of nylon.
$178.30 - $178.30
Chemical Identifiers
| MFCD00001625 |
Specifications
| -47°C | |
| 0.947 g/mL (at 25°C) | |
| 100 mL | |
| 788°F | |
| C6H10(=O) | |
| 3.4 mmHg (20°C) | |
| ≥99.5% (GC); ≥99.5% |
| 108-94-1 | |
| 155°C | |
| 44°C - Closed Cup | |
| Selectophore | |
| MFCD00001625 | |
| 98.14 | |
| For ion-selective electrodes |
Safety and Handling
RTECSNumber : GW1050000
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH