missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Chromoionophore I, MilliporeSigma™ Supelco™

Catalog No. 111012849
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012849 10 mg
111012848 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012849 Supplier Merck Emd Millipore Supplier No. 2708610MGF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chromoionophore I (ETH 5294, 3-octadecanoylimino-7-diethylamino-l,2-benzophenoxazine) is a H+-selective neutral chromoionophore, which is lipophilic in nature and is a highly basic dye molecule.

Chemical Identifiers

MDL Number MFCD00213935
Synonym 3-Octadecanoylimino-7-(diethylamino)-1,2-benzophenoxazine; 9-(Diethylamino)-5-(octadecanoylimino)-5 H-benzo[a]phenoxazine; N-Octadecanoyl-Nile blue; ETH 5294

Specifications

Melting Point 91°C to 93°C
CAS 125829-24-5
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C38H53N3O2
Quantity 10 mg
Formula Weight 583.85
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH