missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Chromoionophore I, MilliporeSigma™ Supelco™
Chromoionophore I (ETH 5294, 3-octadecanoylimino-7-diethylamino-l,2-benzophenoxazine) is a H+-selective neutral chromoionophore, which is lipophilic in nature and is a highly basic dye molecule.
Supplier: Merck Emd Millipore 27086100MGF
Specifications
| 91°C to 93°C | |
| Bottomless Glass Bottle with Inserted Fused Cone | |
| MFCD00213935 | |
| 3-Octadecanoylimino-7-(diethylamino)-1,2-benzophenoxazine; 9-(Diethylamino)-5-(octadecanoylimino)-5 H-benzo[a]phenoxazine; N-Octadecanoyl-Nile blue; ETH 5294 | |
| Selectophore |
| 125829-24-5 | |
| C38H53N3O2 | |
| 100 mg | |
| 583.85 | |
| Selectophore |
Chemical Identifiers
| MFCD00213935 | |
| 3-Octadecanoylimino-7-(diethylamino)-1,2-benzophenoxazine; 9-(Diethylamino)-5-(octadecanoylimino)-5 H-benzo[a]phenoxazine; N-Octadecanoyl-Nile blue; ETH 5294 |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH