missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Chloride ionophore III, MilliporeSigma™ Supelco™

Catalog No. 111018010
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018010 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018010 Supplier Merck Emd Millipore Supplier No. 2489450MG

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00676703
Synonym 3,6-Didodecyloxy-4,5-dimethyl-o-phenylene-bis(mercury chloride); ETH 9033

Specifications

CAS 178959-28-9
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C32H56 Cl2 Hg2O2
Quantity 50 mg
Formula Weight 944.87
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH