missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Chloride ionophore II, Function Tested, MilliporeSigma™ Supelco™

Catalog No. 111018511
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018511 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018511 Supplier Merck Emd Millipore Supplier No. 2490125MG

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00214158
Synonym 4,5-Dimethyl-3,6-dioctyloxy-o-phenylene-bis(mercurytrifluoroacetate); ETH 9009

Specifications

Melting Point 177°C to 179°C
CAS 145889-57-2
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C28H40F6 Hg2O6
Quantity 25 mg
Formula Weight 987.78
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH