missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Cellulose triacetate, MilliporeSigma™ Supelco™

Catalog No. 111015842
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015842 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015842 Supplier Merck Emd Millipore Supplier No. 2219950GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00132680
Synonym Triacetylcellulose

Specifications

CAS 9012-09-3
Solubility Information Chloroform: 0.1 g/10 mL, clear, colorless (leave for 5 h)
Quantity 50 g
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH