missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ Calcium ionophore IV, Function Tested, MilliporeSigma™ Supelco™
Visit our Sensor Applications portal to learn more.
$802.96 - $3117.97
Chemical Identifiers
| MDL Number | MFCD00213934 |
|---|---|
| Synonym | N,N-Dicyclohexyl-N′,N′-dioctadecyl-3-oxapentanediamide; N,N-Dicyclohexyl-N′,N′-dioctadecyl-diglycolic diamide; ETH 5234 |
Chemical Identifiers
| MFCD00213934 | |
| N,N-Dicyclohexyl-N′,N′-dioctadecyl-3-oxapentanediamide; N,N-Dicyclohexyl-N′,N′-dioctadecyl-diglycolic diamide; ETH 5234 |
Specifications
| 50°C to 53°C | |
| Bottomless Glass Bottle with Inserted Fused Cone | |
| MFCD00213934 | |
| N,N-Dicyclohexyl-N′,N′-dioctadecyl-3-oxapentanediamide; N,N-Dicyclohexyl-N′,N′-dioctadecyl-diglycolic diamide; ETH 5234 | |
| Selectophore |
| 126572-74-5 | |
| C52H100N2O3 | |
| 50 mg | |
| 801.36 | |
| Selectophore |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH