missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Cadmium ionophore I, MilliporeSigma™ Supelco™

Catalog No. 111015379
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015379 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015379 Supplier Merck Emd Millipore Supplier No. 2090950MGF

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00143893
Synonym N,N,N′,N′-Tetrabutyl-3,6-dioxaoctanedi(thioamide); ETH 1062

Specifications

Melting Point 34°C to 36°C
CAS 73487-00-0
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C22H44N2O2S2
Quantity 50 mg
Formula Weight 432.73
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH