missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Bis(2-ethylhexyl) sebacate, ≥97.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187901
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012805 5 mL
111012804 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012805 Supplier Merck Emd Millipore Supplier No. 848185ML
Only null left

Bis(2-ethylhexyl) sebacate (BEHS) is a plasticizer widely used in preparing ion-selective electrodes.

Chemical Identifiers

MDL Number MFCD00009497
Synonym Di(2-ethylhexyl) sebacate; Sebacic acid di(2-ethylhexyl) ester; ′Dioctyl′ sebacate

Specifications

Density 0.914 g/mL (at 25°C)
CAS 122-62-3
Boiling Point 212°C (1 mmHg)
Flash Point >113°C
Linear Formula [-(CH2)4 CO2 CH2 CH(C2H5)(CH2)3 CH3]2
Quantity 5 mL
Formula Weight 426.67
Percent Purity ≥97.0% (GC); ≥97.0%
Grade Selectophore
Product Line Selectophore
RTECSNumber VS1000000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH