missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Bis(2-ethylhexyl) adipate, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111012335
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012335 1 mL
111012336 5 mL
111012334 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111012335 Supplier Merck Emd Millipore Supplier No. 021381ML

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00009496
Synonym Adipic acid di(2-ethylhexyl) ester; DOA

Specifications

Melting Point <-70°C
CAS 103-23-1
Density 0.925 g/mL (at 20°C)
Boiling Point 175°C (2 mmHg)
Flash Point 196°C - Closed Cup
Linear Formula [-CH2 CH2 CO2 CH2 CH(C2H5)(CH2)3 CH3]2
Quantity 1 mL
Formula Weight 370.57
Percent Purity ≥99.0% (GC); ≥99.0%
Grade Selectophore
Product Line Selectophore
Show More Show Less
RTECSNumber AU9700000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH