missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Bis(1-butylpentyl)decane-1,10-diyl diglutarate, ≥97%, MilliporeSigma™ Supelco™

Catalog No. 111012286
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012286 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012286 Supplier Merck Emd Millipore Supplier No. 305851G
Only null left

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

MDL Number MFCD00043109
Synonym 5,30-Dibutyl-6,12,23,29-tetraoxatetratriacontane-7,11,24,28-tetrane; ETH 469

Specifications

CAS 101342-76-1
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C38H70O8
Quantity 1 g
Formula Weight 654.96
Percent Purity ≥97%
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH