missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ Ammonium ionophore I, Function Tested (Nonactin, HPLC), ≥95-98.0%, MilliporeSigma™ Supelco™

Catalog No. 111015160
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015160 50 mg
111015159 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015160 Supplier Merck Emd Millipore Supplier No. 0987750MG

Purified for use in ion-selective electrodes

Chemical Identifiers

Synonym Nonactin; Ammonium ionophore

Specifications

CAS 6833-84-7
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C40H64O12
Quantity 50 mg
Formula Weight 736.93
Percent Purity ≥95.0% (Nonactin, HPLC); ≥98.0% (total homologs, HPLC)
Grade Selectophore
Product Line Selectophore
RTECSNumber RH1685000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH