missing translation for 'onlineSavingsMsg'
Learn More

Selectophore™ 1-Tetradecanol, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111018423
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018423 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018423 Supplier Merck Emd Millipore Supplier No. 871585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Visit our Sensor Applications portal to learn more.

Chemical Identifiers

CAS 112-72-1
Molecular Weight (g/mol) 214.39
MDL Number MFCD00004757
Synonym Myristyl alcohol; Tetradecyl alcohol

Specifications

Melting Point 35°C to 39°C; 38°C to 39°C
Density 0.823 g/mL (at 25°C)
Boiling Point 289°C
Flash Point 148°C
Ignition Point 489°F
Linear Formula CH3(CH2)13 OH
Quantity 5 g
Vapor Density 7.4 (vs. air)
Formula Weight 214.39
Percent Purity ≥99.0% (GC); ≥99.0%
Grade Selectophore
Physical Form Powder
Product Line Selectophore
Chemical Name or Material 1-Tetradecanol
Show More Show Less
RTECSNumber XB8655000

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH