missing translation for 'onlineSavingsMsg'
Learn More

SEKISUI Diagnostics™ OSOM™ Ultra Strep A Test

Catalog No. 22071067
Change view
Click to view available options
Quantity:
25 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22071067 25 Tests
22071068 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22071067 Supplier SEKISUI Diagnostics™ Supplier No. SEK 147

Includes

Controls

The OSOM™ Ultra Strep A Test offers faster results for faster treatment—statistically equivalent to single swab culture with results in just five minutes.

  • Simple-to-use - get easy to read results in four easy steps and no need for additional equipment
  • Greater sensitivity - antibody labeled color particles coated at two separate locations allows for greater sensitivity than conventional single label technology without sacrificing specificity
  • Easy-to-read two-color results
  • Simplify reagent addition - single reagent delivery system in 25-test kit eliminates the need to handle two separate reagents
  • QC Inside™ - two extra tests included in the 50-test kit at no additional charge to run external quality control for more confidence in your results
  • Made in the USA

Specifications

Accuracy 92.6%
Content And Storage Room temperature (15°C to 30°C, 59°F to 86°F)
Control Sets Yes, included
CPT Code 87880QW
Clia Complexity Waived
Detectable Analytes Streptococci (Group A) Antigen
DoA Calibrators No
Format Manual Dipstick
Sample Type Throat Swab
Sensitivity 96.4% (95% CI: 93.0% to 97.9%)
Type Rapid Test
For Use With (Application) Lateral Flow Immunoassay
Includes 25 test sticks, 25 test tubes, 25 sterile swabs, 25 extraction reagent bottles, 1 positive control, 1 negative control, 1 workstation, and 1 directional insert
Packaging Type Test Kit
Quantity 25 Tests
Specificity 96.3% (95% CI: 94.3% to 97.5%)
Test Time 5 min.
Analysis Time 5 min.
Show More Show Less