missing translation for 'onlineSavingsMsg'
Learn More

SEKISUI Diagnostics™ OSOM™ Trichomonas Rapid Test

Catalog No. 22071071
Change view
Click to view available options
Type:
Control Kit
Rapid Test
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22071071 Rapid Test
22071072 Control Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22071071 Supplier SEKISUI Diagnostics™ Supplier No. SEK 181
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The OSOM™ Trichomonas Rapid Test is the only CLIA-waived lateral flow rapid test for the detection of Trichomoniasis while providing 95% agreement with culture and wet mount combined.

  • Simple-to-use - get easy to read results in just ten minutes and four easy steps
  • Eliminate the need for wet mount - antigen-based rapid test does not require visualization of live organisms
  • CDC-approved method of testing for the detection of T. vaginalis - unaffected by other common infections
  • Trust your results - 95% agreement against Composite Reference Standard (culture and wet mount)
  • Easy-to-use - requires minimal training and no additional equipment
  • QC Inside™ - two extra tests included at no additional charge to run external quality control testing for more confidence in your results
  • Made in the USA

Specifications

Content And Storage Room temperature (15°C to 30°C, 59°F to 86°F)
CPT Code 87808QW
Clia Complexity Waived
Detectable Analytes T. vaginalis Antigens
Sensitivity 96% (95% CI, 91% to 100%)
Type Rapid Test
For Use With (Application) Lateral Flow Immunoassay
Includes 25 test sticks, 25 sterile swabs, 25 test tubes, 1 sample buffer vial, 1 sample buffer dropper top, 1 positive control swab, 1 workstation, and 1 instructions for use (IFU)
Specificity 95% (95% CI, 92% to 97%)
Test Time 10 min.
Analysis Time 10 min.
Show More Show Less