missing translation for 'onlineSavingsMsg'
Learn More

SEKISUI Diagnostics™ OSOM™ Mono Test Kit

Catalog No. 22071069
Change view
Click to view available options
Analysis Time:
5 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Analysis Time
22071069 5 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22071069 Supplier SEKISUI Diagnostics™ Supplier No. SEK 145
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

OSOM™ Mono Test Kit for simple, in-office test for the qualitative detection of infectious mononucleosis with no age limitation on patients that can be tested.

  • One test, all ages - no age limit on who can be tested
  • Simple-to-use - CLIA-waived (whole blood) with easy-to-read results in just three steps, with no need for additional equipment
  • Trust your results - high sensitivity and specificity with results in just 5 minutes
  • Easy-to-read two-color results - blue test line and red control line
  • QC Inside™ - two extra tests and controls included at no additional charge to run external quality control testing for more confidence in your results

Specifications

Content And Storage Room Temperature (15°C to 30°C, 59°F to 86°F)
Control Sets Included
CPT Code 86308QW (Whole Blood), 86308 (Serum or Plasma)
Clia Complexity Waived (Whole Blood)
Detectable Analytes Mononucleosis Heterophil Antibodies
Format Manual Dipstick
Sample Type Serum, Plasma, or Whole Blood
Sensitivity >99%
Type Rapid Test
Includes 25 test sticks, 25 test tubes, 25 transfer pipettes, 25 capillary pipettes, 1 diluent, 1 Mono positive control, 1 Mono negative control, 1 workstation, 1 instructions for use (IFU), full-color pictorial procedure for convenient reference, 2 additional test sticks are included for external QC testing
Packaging Type Test Kit
Specificity 96%
Test Time 5 min.
Analysis Time 5 min.
Show More Show Less