missing translation for 'onlineSavingsMsg'
Learn More

SEKISUI Diagnostics™ OSOM™ hCG Urine Controls

Catalog No. 22071066
Change view
Click to view available options
For Use With (Equipment):
OSOM hCG Test Kits
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22071066 OSOM hCG Test Kits
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22071066 Supplier SEKISUI Diagnostics™ Supplier No. SEK 134

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Meet your quality control requirements of your hCG testing with this set of OSOM™ hCG Urine Controls. Contains positive and negative hCG urine controls.

For use with:

  • OSOM™ hCG Urine Test Kit (SEKISUI Diagnostics part number 101)
  • OSOM™ hCG Card Pregnancy Test Kit (SEKISUI Diagnostics part number 102)
  • OSOM™ hCG Combo Test Kit (SEKISUI Diagnostics part number 124)
  • OSOM™ Ultra hCG Combo Test Kit (SEKISUI Diagnostics part number 1004)

Specifications

Content And Storage 2°C to 8°C (35.6°F to 46.4°F)
For Use With (Equipment) OSOM hCG Test Kits
Sample Type Urine
Type Control Kit
Includes 10 mL vial of negative urine control, 10 mL vial of positive urine control
Intended Use Quality control testing with OSOM hCG-Urine (101), OSOM Card Pregnancy (102), and OSOM hCG Combo (124) Tests