missing translation for 'onlineSavingsMsg'
Learn More

SEKISUI Diagnostics™ OSOM™ hCG Serum Control Set

Catalog No. 22071091
Change view
Click to view available options
For Use With (Equipment):
OSOM hCG Combo Test Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22071091 OSOM hCG Combo Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22071091 Supplier SEKISUI Diagnostics™ Supplier No. SEK 138
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

One 5mL vial hCG Negative Serum Control and one 5mL vial hCG Positive Serum Control

Meet your quality control requirements of your hCG testing with this OSOM™ hCG Serum Control Set.

  • Contains positive and negative hCG serum controls
  • For use with OSOM™ hCG Combo Test (SEKISUI Diagnostics part numbers 124 and 1004)

Specifications

Content And Storage 2°C to 8°C (35.6°F to 46.4°F)
For Use With (Equipment) OSOM hCG Combo Test Kit
Sample Type Whole Blood, Serum, Plasma, Urine
Type Control Kit
Includes One 5 mL vial hCG Negative Serum Control, One 5 mL vial hCG Positive Serum Control
Intended Use Quality control testing with the OSOM hCG Combo Test (124)