Learn More
3M™ SecureFit™ 600 Series Protective Eyewear
3M™ SecureFit™ 600 Series Protective Eyewear features proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
Supplier:  3M™ 7100132255
 View more versions of this product
Catalog No. 17-000-695
Only null left
Explore available promotions
Description
Description
  • Scotchgard™ Anti-fog Coating lasts longer than traditional anti-fog coatings, helping to increase eyewear performance in wet and steamy environments
  • The proprietary coating retains its effectiveness for at least 25 washings, allowing workers to wear their eyewear longer
  • Workers can feel confident wearing Scotchgard™ Anti-Fog coated eyewear because it meets the requirements of ANSI Z87.1-2015
  • Rugged Anti-Scratch (RAS) coating helps extend the life of the lens
  • Indoor/Outdoor Grey lens helps reduce brightness and glare when working both indoors and outdoors
  • Grey lens are popular in outdoor daytime work environments
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated spectacle
  • Lens absorbs 99.9% UVA & UVB rays
  • 3M Catalog# SF601RAS, SF602RAS, SF603SGAF, SF607SGAF, SF613AS, SF617AS, SF630AS
Specifications
Specifications
Scratch-resistant, UV-treated
Shade 3
Gray
20/Pk.
Polycarbonate
UV400
ANSI Z87.1-2015
Safety Glasses
SF630AS
SecureFit™
Plastic
Half Frame
Product Suggestions
Product Suggestions