Learn More
3M™ SecureFit™ 400X Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
3M™ SecureFit™ 400X Safety Glasses have 3M™ Pressure Diffusion Temple Technology, flexible temple arms and combined soft nose bridge/brow guard. Lightweight anti-scratch & superior anti-fog, rimless polycarbonate lens for indoor/outdoor use.
$284.00 - $284.00
Specifications
| Certifications/Compliance | CE Approved |
|---|---|
| Temple Style | Pressure Diffusion Temple Technology |
| Lens Material | Polycarbonate |
| Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity | ||||
|
296367922
|
3M™
SF401XSGAFBLU |
Blue, Gray | Clear |
Case of 20 Each for $284.00
|
|
||||
|
296367925
|
3M™
SF402XSGAFBLU |
Blue, Gray | Gray |
Case of 20 Each for $284.00
|
|
||||
|
296367924
|
3M™
SF407XSGAFBLU |
Blue, Gray | Light Gray, Indoor/Outdoor |
Case of 20 Each for $284.00
|
|
||||
Description
- Safety glasses with 3M™ Pressure Diffusion Temple Technology for comfort
- Wraparound design to improve the security of fit and prevent slipping
- Padded temple touchpoints for increased comfort over the ears
- Soft nose bridge for a more comfortable fit
- Contoured brow guard provides additional coverage from flying particles
- Superior anti-fog and anti-scratch properties meets the K&N marking requirements of EN166
- Frame marking: SF400 Series, 3M EN166 FT
Specifications
| CE Approved | |
| Pressure Diffusion Temple Technology | |
| Polycarbonate |