missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SecurAlert Self-Expiring Time Badges

Catalog No. p-6768137
Change view
Click to view available options
For Use With (Application):
Contractors
Visitors
Product Type:
One day expiration
One week expiration
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Product Type
19103182 Visitors One day expiration
19103183 Contractors One day expiration
19103176 Contractors One week expiration
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19103182 Supplier Brady™ Supplier No. 95688
Only null left

Minimize unauthorized use of visitor badges. Brady™ SecurAlert Self-Expiring Time Badges offer a clear indication of a visitor's authorization.

  • Automatically expire to prevent unauthorized use
  • Once the "front" is placed on the "back" badge, the expiration process is started and cannot be reversed
  • Easy-to-see color change confirms whether or not a visitor is authorized to be on your premises
  • Self-adhesive back for easy application

Specifications

Length (English) 2 in.
Color Red on white
Dimensions (L x W) 3 in. x 2 in.
For Use With (Application) Visitors
Width (English) 3 in.
Product Type One day expiration