missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ SecurAlert Clip-On Badges
Description
- Self-expiring badges
- Low cost clip-on alternative to self-adhesive badges
- 3 in. x 3 in. slotted cards
- 500 front and back badge pairs per pack
Specifications
Specifications
| Item Description | 1 week expiration |
| Length (English) | 3 in. |
| Color | Red |
| Dimensions (L x W) | 3 in. x 3 in. |
| For Use With (Application) | Visitor |
| Width (English) | 3 in. |