missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SecurAlert Clip-On Badges

Catalog No. 19103150
Change view
Click to view available options
For Use With (Application):
Contractor
Temporary
Visitor
Item Description:
1 day expiration
1 week expiration
Color:
Blue
Green
Red
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Item Description Color
19103150 Visitor 1 week expiration Red
19103153 Visitor 1 week expiration Green
19103154 Visitor 1 week expiration White
19103163 Contractor 1 week expiration Green
19103165 Temporary 1 day expiration Red
19103169 Temporary 1 day expiration White
19103170 Temporary 1 week expiration Red
19103174 Temporary 1 week expiration White
19103166 Temporary 1 day expiration Blue
19103167 Temporary 1 day expiration Yellow
19103173 Temporary 1 day expiration Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19103150 Supplier Brady™ Supplier No. 95648

Prevent unauthorized use of visitor, contractor and temporary passes. Brady™ SecurAlert Clip-On Badges are self-expiring, which prevents unauthorized use beyond their intended use periods.

  • Self-expiring badges
  • Low cost clip-on alternative to self-adhesive badges
  • 3 in. x 3 in. slotted cards
  • 500 front and back badge pairs per pack

Specifications

Item Description 1 week expiration
Length (English) 3 in.
Color Red
Dimensions (L x W) 3 in. x 3 in.
For Use With (Application) Visitor
Width (English) 3 in.